Nonadecane 99%
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Usage: Organic building block/reagent.
Synonymer: N-nonadecane
- CAS nummer:629-92-5
- Formel:C₁₉H₄₀
- MDL nummer:MFCD00009012
- Molecular Weight:268,52 g/mol
5 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Organic building block/reagent.
Synonymer: N-nonadecane
- CAS nummer:629-92-5
- Formel:C₁₉H₄₀
- MDL nummer:MFCD00009012
- Molecular Weight:268,52 g/mol
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



