Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Glutamic acid ≥98%

L-Glutamic acid ≥98%

Bottle
Katalog # APOSBUP19804-5G
New Product
Leverandør:   Apollo Scientific
L-Glutamic acid ≥98%
Katalog # APOSBUP19804-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  56-86-0
L-Glutamic acid ≥98%

Bottle

CAS nummer:  56-86-0
Price
407,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS nummer:56-86-0
  • Formel:C₅H₉NO₄
  • Molecular Weight:147,13 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.