MES monohydrate ≥98%
Bottle
Katalog # APOSBUP19522-100G
New Product
Leverandør: Apollo Scientific
MES monohydrate ≥98%
Katalog # APOSBUP19522-100G
Leverandør: Apollo Scientific
New Product
CAS nummer: 145224-94-8
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
- CAS nummer:145224-94-8
- Formel:C₆H₁₅NO₅S
- Molecular Weight:213,252 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



