Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

DTNB ≥95%

Irriterende
New Product
Leverandør:   Apollo Scientific
DTNB ≥95%
Leverandør:  Apollo Scientific
New Product
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS nummer:69-78-3
  • Formel:C₁₄H₈N₂O₈S₂
  • Molecular Weight:396,35 g/mol
  • EINECS:200-714-4
Product Family Options

Produktinformation

  • Pakning
    Tilgængelighed
    Price
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 g
    1 * 1 g - 286,00 kr
    286,00 kr
    1 * 1 g
       286,00 kr

    Specifikationer:
    New Product
    New Product
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 g
    1 * 5 g - 505,00 kr
    505,00 kr
    1 * 5 g
       505,00 kr

    Specifikationer:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.