Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Eugenol ≥98%

Bottle
  APOSBUP18059-500MG
New Product
Eugenol ≥98%
  APOSBUP18059-500MG
New Product
 :  97-53-0
Eugenol ≥98%

Bottle

 :  97-53-0
 
286,00 kr
   
1 * 500 mg
 

Usage: Biochemical/life science applications.

 :  4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
  •  :97-53-0
  •  :C₁₀H₁₂O₂
  •  :164,2 g/mol
 

 

  • Pakning:
    Bottle