Finasteride ≥98%
New Product
Supplier: Apollo Scientific
Gå til:
Om denne vare
Usage: Biochemical/life science applications.
Synonymer: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
- CAS nummer:98319-26-7
- Formel:C₂₃H₃₆N₂O₂
- Molecular Weight:372,54 g/mol
4 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
- CAS nummer:98319-26-7
- Formel:C₂₃H₃₆N₂O₂
- Molecular Weight:372,54 g/mol
Product Family Options
Produktinformation
- PakningAvailabilityPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product



