Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinc Borate, anhydrous 99% 5 - 9 μm

Zinc Borate, anhydrous 99% 5 - 9 μm

New Product
Leverandør:   Apollo Scientific
Zinc Borate, anhydrous 99% 5 - 9 μm
Leverandør:  Apollo Scientific
New Product
Certifikater
Zinc Borate, anhydrous 99% 5 - 9 μm

2 tilgængelige muligheder nedenfor

Product Details & Documents

Usage: Inorganic salt or catalyst

Synonymer:  Diboron zinc tetraoxide
  • CAS nummer:1332-07-6
  • Formel:B₂O₆Zn₃
  • Molecular Weight:313,76 g/mol
Product Family Options

Produktinformation

  • Pakning
    Tilgængelighed
    Price
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 g
    1 * 100 g - 165,00 kr
    165,00 kr
    1 * 100 g
       165,00 kr

    Specifikationer:
    New Product
    New Product
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 g
    1 * 500 g - 275,00 kr
    275,00 kr
    1 * 500 g
       275,00 kr

    Specifikationer:
    New Product
    New Product
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.