Orders LinkContactUsLinkComponent
Methyl Oleate ≥98%
Bottle
  APOSBUP18981-1G
New Product
Methyl Oleate ≥98%
  APOSBUP18981-1G
New Product
 :  112-62-9
Methyl Oleate ≥98%
Bottle
 :  112-62-9
297,00 kr
1 * 1 g
Usage: Biochemical/life science applications
 :  cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
  •  :112-62-9
  •  :C₁₉H₃₆O₂
  •  :296,49 g/mol
  • Pakning:
    Bottle