Fenofibrate ≥98%
New Product
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Usage: Biochemical/life science applications
Synonymer: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
- CAS nummer:49562-28-9
- Formel:C₂₀H₂₁ClO₄
- Molecular Weight:360,83 g/mol
2 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Biochemical/life science applications
Synonymer: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
- CAS nummer:49562-28-9
- Formel:C₂₀H₂₁ClO₄
- Molecular Weight:360,83 g/mol



