Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Gemfibrozil ≥98%

Gemfibrozil ≥98%

New Product
Supplier:   Apollo Scientific
Gemfibrozil ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Gemfibrozil ≥98%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  CI-719
  • CAS Number:25812-30-0
  • Formula:C₁₅H₂₂O₃
  • Molecular Weight:250,33 g/mol
Product Family Options

Product Information

  • Pakning
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 483,00 kr
    483,00 kr
    1 * 100 mg
       483,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 374,00 kr
    374,00 kr
    1 * 50 mg
       374,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 615,00 kr
    615,00 kr
    1 * 200 mg
       615,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 3.180,00 kr
    3.180,00 kr
    1 * 5 g
       3.180,00 kr

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.