Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Alanine 98%

L-Alanine 98%

Bottle
Catalog # APOSBI9899-1KG
New Product
Supplier:   Apollo Scientific
L-Alanine 98%
Catalog # APOSBI9899-1KG
Supplier:  Apollo Scientific
New Product
CAS Number:  56-41-7
L-Alanine 98%

Bottle

CAS Number:  56-41-7
Price
516,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents

Usage: Biochemical/life science applications

  • Non-hazardous
Synonyms:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS Number:56-41-7
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00064410
  • Molecular Weight:89,093 g/mol
  • EINECS:200-273-8
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.