Eosin B biological stain
New Product
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Usage: Organic building block/reagent
Synonymer: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
- CAS nummer:548-24-3
- Formel:C₂₀H₆Br₂N₂Na₂O₉
- MDL nummer:MFCD00005041
- Molecular Weight:624,06 g/mol
5 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Organic building block/reagent
Synonymer: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
- CAS nummer:548-24-3
- Formel:C₂₀H₆Br₂N₂Na₂O₉
- MDL nummer:MFCD00005041
- Molecular Weight:624,06 g/mol
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product



