Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HEPES ≥98%

HEPES ≥98%

Bottle
Catalog # APOSBUP18543-500MG
New Product
Supplier:   Apollo Scientific
HEPES ≥98%
Catalog # APOSBUP18543-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  7365-45-9
HEPES ≥98%

Bottle

CAS Number:  7365-45-9
Price
396,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  • CAS Number:7365-45-9
  • Formula:C₈H₁₈N₂O₄S
  • Molecular Weight:238,3 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications