Metanilyellow indicator
New Product
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Usage: Organic building block/ reagent.
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375,38 g/mol
6 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Organic building block/ reagent.
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375,38 g/mol
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



