Dibutyl phthalate ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: DBP
- CAS Number:84-74-2
- Formula:C₁₆H₂₂O₄
- Molecular Weight:278,34 g/mol
2 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: DBP
- CAS Number:84-74-2
- Formula:C₁₆H₂₂O₄
- Molecular Weight:278,34 g/mol



