Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Hydroxyflavanone ≥98%

2-Hydroxyflavanone ≥98%

New Product
Leverandør:   Apollo Scientific
2-Hydroxyflavanone ≥98%
Leverandør:  Apollo Scientific
New Product
Certifikater
2-Hydroxyflavanone ≥98%

2 tilgængelige muligheder nedenfor

Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS nummer:17348-76-4
  • Formel:C₁₅H₁₂O₃
  • Molecular Weight:240,25 g/mol
Product Family Options

Produktinformation

  • Pakning
    Tilgængelighed
    Price
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mg
    1 * 100 mg - 297,00 kr
    297,00 kr
    1 * 100 mg
       297,00 kr

    Specifikationer:
    New Product
    New Product
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 mg
    1 * 200 mg - 439,00 kr
    439,00 kr
    1 * 200 mg
       439,00 kr

    Specifikationer:
    New Product
    New Product
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.