Lids for All-Purpose Storage Containers
Product Details & Documents
Precisely fitting protection for storage, logistics and transportation.
Particularly robust, durable and resistant to chemicals and mechanical stress
- Reliably protects against external influences such as dust, moisture and dirt
- Easy handling thanks to support construction
- Dimensionally stable and stackable, even with intensive use
The support lid is the perfect addition to Bürkle all-purpose storage containers. Made from high-quality HDPE, the lid impresses with its robustness, durability and precise fit. The lid can be fitted effortlessly and removed just as quickly.
In daily use, the lid offers reliable protection against dust, dirt and mechanical influences. The plastic lid has proven itself as a functional accessory for transportation, storage and internal material flow, particularly in laboratories, industry and trade. Even under heavy use, the material remains dimensionally stable and meets the highest standards of hygiene, safety and efficiency.
Specifikationer
- Farve:hvid
- L×B:400×300 mm
- Materiale:HDPE