InfinityLab 96-well Plate
Product Details & Documents
This storage plate was designed for a wide variety of chromatography applications and for sample collection, storage, precipitation, and centrifugation. The V-shaped bottoms provide excellent small-volume recovery with autosamplers.
Clear, flat for uniform sealing
Alphanumeric grid; Autoclavable
Stackable, SBS footprint
Made from highest medical grade virgin polypropylene
Very low biomolecular binding characteristics
Resistance to many standard laboratory chemicals, including DMSO, phenol, and chloroform
Free from RNase, DNase, human genomic DNA, and endotoxins
Suitable for chromatography
High temperature tolerance: –80 to 120 °C
Specifikationer
- Anbefalet arbejdsvolumen:300 µl
- Steril:-
- Farve:klar
- Beskrivelse:InfinityLab 96-well plate, well shape: round, V shape bottom, 14 mm
- Brøndvolumen:330 µl
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Closing mats for 96 well plates, silicone, pre-slit, round wells, compatible with AGLS5043-9313Closing mats for 96 well plates, silicone, pre-slit, round wells, compatible with AGLS5043-9313AGLS5042-1389Log ind for tilgængelighed1 * 50 Items
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



