Orders LinkContactUsLinkComponent
Stainless Steel Double-Ended Spatulas
New Product
Leverandør:   BURKLE
Stainless Steel Double-Ended Spatulas
Leverandør:  BURKLE
New Product
Certifikater
Stainless Steel Double-Ended Spatulas
9 tilgængelige muligheder nedenfor
Product Details & Documents
Sturdy spatulas for precise dosing and gentle spreading.
  • Two edges: angular for precise work, rounded (curved) for gentle picking up and spreading
  • Made of high-quality stainless steel for durability, hygiene, and corrosion resistance
  • Versatile use; ideal for dosing, picking up, spreading, stirring, or mixing
This high-quality stainless steel double spatula is the ideal tool for precise and versatile everyday laboratory work. The spatula has a flat, angular edge on one side and a rounded (flat or curved) edge on the other. Whether portioning fine samples precisely, removing substances gently, or distributing larger quantities evenly, this spatula is suitable for every task. Thanks to its sophisticated geometry and solid stainless steel 1.4301 (304) construction, it is durable, hygienic and corrosion-resistant. An ideal tool for demanding applications in laboratories and industry.
The square edge is perfect for scraping, cutting or portioning solid or powdery substances, especially in the corners of cans or flat containers. The rounded side is ideal for removing, distributing, or stirring substances in round containers without scratching them. This double spatula replaces several individual tools, saving time and increasing work safety while reducing the risk of cross-contamination.
 : Spatulas are packaged individually.
Product Family Options
Produktinformation
  • Længde
    Klinge længde
    Bladbredde
    Farve
    Tilgængelighed
    Price
  • BURK5386-0080
    Chattaway double-ended stainless steel spatula with square/round curved tip
    Chattaway double-ended stainless steel spatula with square/round curved tip
    125 mm
    40 mm
    8 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 18,60 kr
    18,60 kr
    1 * 1 Items
       18,60 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0081
    Chattaway double-ended stainless steel spatula with square/round curved tip
    Chattaway double-ended stainless steel spatula with square/round curved tip
    150 mm
    45 mm
    8 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 19,40 kr
    19,40 kr
    1 * 1 Items
       19,40 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0082
    Chattaway double-ended stainless steel spatula with square/round curved tip
    Chattaway double-ended stainless steel spatula with square/round curved tip
    184 mm
    50 mm
    10 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 22,10 kr
    22,10 kr
    1 * 1 Items
       22,10 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0083
    Chattaway double-ended stainless steel spatula with square/round curved tip
    Chattaway double-ended stainless steel spatula with square/round curved tip
    200 mm
    63 mm
    10 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 23,70 kr
    23,70 kr
    1 * 1 Items
       23,70 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0084
    Chattaway double-ended stainless steel spatula with square/round curved tip
    Chattaway double-ended stainless steel spatula with square/round curved tip
    237 mm
    66 mm
    11 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 27,50 kr
    27,50 kr
    1 * 1 Items
       27,50 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0041
    Double-ended stainless steel spatula with angular/rounded tip
    Double-ended stainless steel spatula with angular/rounded tip
    150 mm
    45 mm
    22 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 17,10 kr
    17,10 kr
    1 * 1 Items
       17,10 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0043
    Double-ended stainless steel spatula with angular/rounded tip
    Double-ended stainless steel spatula with angular/rounded tip
    210 mm
    60 mm
    22 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 28,00 kr
    28,00 kr
    1 * 1 Items
       28,00 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0045
    Double-ended stainless steel spatula with angular/rounded tip
    Double-ended stainless steel spatula with angular/rounded tip
    300 mm
    75 mm
    22 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 99,90 kr
    99,90 kr
    1 * 1 Items
       99,90 kr
    Specifikationer:
    New Product
    New Product
  • BURK5386-0048
    Double-ended stainless steel spatula with angular/rounded tip
    Double-ended stainless steel spatula with angular/rounded tip
    500 mm
    100 mm
    22 mm
    sølvfarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 161,00 kr
    161,00 kr
    1 * 1 Items
       161,00 kr
    Specifikationer:
    New Product
    New Product