TapeStation Platform Compliance Solution
About this item
The Agilent tapestations compliance solution provides an integrated and reliable tool for quality control of nucleic acids. It supports the validation process from start to finish, enabling users to run the Agilent tapestation systems in regulated lab environments. The Agilent tapestation compliance solution comprises the 4150 and 4200 tapestation instrument, software, services, as well as application-specific consumables.
Reproducible results: System with prepackaged reagents and screentape devices, standardized protocols and optimized assays
Objective information regarding sample concentration, size and integrity
Compliance services: Software and hardware IQ and OQ can be purchased at any time. A certified engineer will test and verify the functionality of the hardware and software, thereby qualifying the system
Design qualification: Functional and operational specifications are defined by the declaration of conformity for the tapestation instruments, screentape devices as well as the tapestation test tape, certificate of conformance for the reagents and the declaration of system validation for the Agilent tapestation software
Certifications: GLP/GMP compliant.
2 Options Available Below
Product Details & Documents
The Agilent tapestations compliance solution provides an integrated and reliable tool for quality control of nucleic acids. It supports the validation process from start to finish, enabling users to run the Agilent tapestation systems in regulated lab environments. The Agilent tapestation compliance solution comprises the 4150 and 4200 tapestation instrument, software, services, as well as application-specific consumables.
Reproducible results: System with prepackaged reagents and screentape devices, standardized protocols and optimized assays
Objective information regarding sample concentration, size and integrity
Compliance services: Software and hardware IQ and OQ can be purchased at any time. A certified engineer will test and verify the functionality of the hardware and software, thereby qualifying the system
Design qualification: Functional and operational specifications are defined by the declaration of conformity for the tapestation instruments, screentape devices as well as the tapestation test tape, certificate of conformance for the reagents and the declaration of system validation for the Agilent tapestation software
Certifications: GLP/GMP compliant.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- AGLSSYS-TS-4150
- AGLSSYS-TS-4200
- DNA:
- Yes
- Yes
Specifications
Product Family Options
Product Information
- BeskrivelseAvailabilityPrice
- Specifications:New Product
More
Specifications
Cat. No.AGLSSYS-TS-4150DNAYesNew Product - Specifications:New Product
More
Specifications
Cat. No.AGLSSYS-TS-4200DNAYesNew Product