Blocks for COD
New Product
Leverandør: VELP SCIENTIFIC
Gå til:
Om denne vare
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
3 tilgængelige muligheder nedenfor
Product Details & Documents
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Product Family Options
Produktinformation
- BrøndstørrelseTilAnt. hullerDybdeTilgængelighedPrice
- 705-1207COD-W04-D420-A blockCOD-W04-D420-A blockØ 42,0 mm42 mm round-bottom tubes451 mmLog ind for tilgængelighed1 * 1 Items915,00 krSpecifikationer:New ProductNew Product
- 705-1208COD-W20-COMBO blockCOD-W20-COMBO blockØ 16,0/22,0 mm16/22 mm round-bottom tubes18/247/45,5 mmLog ind for tilgængelighed1 * 1 Items915,00 krSpecifikationer:New ProductNew Product
- 705-1206COD-W24-D160-A blockCOD-W24-D160-A blockØ 16,0 mm16 mm round-bottom tubes2447 mmLog ind for tilgængelighed1 * 1 Items915,00 krSpecifikationer:New ProductNew Product