Ubbelohde Viscometer (DIN)
Ubbelohde viscometer (DIN) with TC sensors, K = 0,3, 10...80 °C
Catalog # SCOI285423475
New Product
Supplier: SI Analytics
Jump to:
Product Details & Documents
Ubbelohde viscometer (DIN) with TC sensors, 587 23, calibrated.
Calibrated for automatic measurements, capillary no. IIc, constant K = 0,3.
Specifications
- Temperaturområde:10...80 °C
- Beskrivelse:Ubbelohde viscometer (DIN) with TC sensors
- Konstant:K = 0,3