Orders LinkContactUsLinkComponent
Test Tubes with Screw Cap
Test Tubes with Screw Cap
New Product
Supplier:   KARTELL
Test Tubes with Screw Cap
Supplier:  KARTELL
New Product
Certificates
Test Tubes with Screw Cap
3 Options Available Below
Product Details & Documents
PP test tubes, 10 ml, with self-standing base and tight screw cap yellow colour for a leakproof closure.
  • Not graduated
    Available also in sterile and individually wrapped version
    Ø×H: 16×100 mm
Product Family Options
Product Information
  • Kapacitet
    Kasket inkluderet
    Lukkestørrelse
    Beskrivelse
    Availability
    Price
  • 10 ml
    Yes
    Skruelåg
    Test tubes, non sterile
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items
    Specifications:
    New Product
    New Product
  • 10 ml
    Yes
    Skruelåg
    Test tubes, sterile individually wrapped
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items
    Specifications:
    New Product
    New Product
  • 10 ml
    Yes
    Skruelåg
    Test tubes, sterile
    Log in for availability
    Log in for availability
    1 * 1.000 Items
    1 * 1.000 Items
    Specifications:
    New Product
    New Product