iQue® Plus One Detector Maintenance Solution
Product Details & Documents
The iQue® Plus One Detector Maintenance Solution aids in rinsing flow cytometry equipment by acting as a lubricant, allowing broken down debris to pass more easily. Typically, this is used during startup/shutdown cycles and during internal cleanings.
Convenient, ready-to-use solution
Second phase rinsing solution for cleaning the detector - prevents clogging
Applications: Maintenance for (500 ml, 1X concentrate - 5 bottles) for iQue® 3 and iQue® Screener Plus
iQue® Plus One Detector Maintenance Solution has revolutionized the flow cytometry maintenance process. This ready-to-use solution prevents clogging of equipment and acts as a lubricating agent to assist in the passing of broken-down debris.
Device/Instrument compatibility: iQue® Screener PLUS and iQue® 3
Specifikationer
- Beskrivelse:iQue® Plus One detector maintenance solution, 2500 ml