Blocks for Vials
New Product
Leverandør: VELP SCIENTIFIC
Gå til:
Om denne vare
These blocks are suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
9 tilgængelige muligheder nedenfor
Product Details & Documents
These blocks are suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Product Family Options
Produktinformation
- BrøndstørrelseTilAnt. hullerDybdeTilgængelighedPrice
- VELPA00000514VLS-W20-D120-A blockSpecifikationer:New ProductNew Product
- VELPA00000519VLS-W09-D210-A blockSpecifikationer:New ProductNew Product
- VELPA00000520VLS-W08-D230-A blockSpecifikationer:New ProductNew Product
- VELPA00000521VLS-W08-D250-A blockSpecifikationer:New ProductNew Product
- VELPA00000516VLS-W15-D160-A blockSpecifikationer:New ProductNew Product
- VELPA00000517VLS-W12-D170-A blockSpecifikationer:New ProductNew Product
- VELPA00000515VLS-W20-D150-A blockSpecifikationer:New ProductNew Product
- VELPA00000518VLS-W12-D190-A blockSpecifikationer:New ProductNew Product
- VELPA00000522VLS-W06-D280-A blockSpecifikationer:New ProductNew Product