Handheld Digital Microscope, Edge AF7515MT4A
Product Details & Documents
Integrated with coaxial lighting for viewing reflective surfaces, AF7515MT4A is a 5-megapixel microscope delivering exceptional brightfield imaging at 420x ~ 470x magnification. Through the use of FLC, this model can offer clear visualization of different topographical features such as edges, dents, and pores. AF7515MT4A is also compatible with the optional WF-20 Wi-Fi streamer, providing users with added convenience for wireless streaming applications.
High Optical Resolution: The superior optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications.
5.0 Megapixel: The 5-megapixel sensor excels at capturing fine details and can transmit high-quality images at a resolution of 2592 x 1944.
Coaxial-Brightfield Illumination: The coaxial illumination uses a through-the-lens design, offering a compact yet high-quality solution for revealing imperfections and fine details on reflective surfaces.
Flexible LED Control (FLC, coaxial light): Tasking with software, FLC provides a 6-level intensity control for the activated LED quadrants and the coaxial light.
Automatic Magnification Readout (AMR):
The Automatic Magnification Readout (AMR) detects the magnification in use, eliminating manual input errors and enhancing measurement accuracy in tasks with frequent magnification adjustments.
Adaptable Interface: The adaptable interface provides the capability of transforming Dino-Lite into a wireless microscope when attaching with the WF-20.
Robust Metal Housing: The anodized aluminum alloy housing offers durability and EMI shielding, effectively guarding against impacts, abrasions, and electromagnetic noise.
Specifikationer
- Forstørrelse:420x ~ 470x
- Model:AF7515MT4A
- Cat. No.:IDCPAF7515MT4A
- Opløsning:5.0 Megapixel (2592×1944)
- Vægt:Approx. 101 g
- Software:DinoCapture 2.0, DinoCapture 3.0, DinoXcope