LiveONLY™ Nuclear Red
LiveONLY™ Nuclear Red is a nuclear dye designed to stain the nucleus exclusively in live cells. This unique feature allows for clear distinction between live cells and dead or membrane-compromised cells. Its cell-permeable design enables selective accumulation in the nucleus, producing red fluorescence without requiring fixation or permeabilization. The simple mix-and-read protocol requires minimal hands-on time, making it ideal for seamless integration into standard imaging workflows. Compatible with a wide range of cell lines and imaging systems, LiveONLY™ Nuclear Red is ideal for live-cell applications such as real-time cell tracking, viability assessment, and high-content screening.
Solvent: DMSO (Dimethyl sulfoxide)
H‑phrase: H303, H313, H333
- Hazard symbol: XN
- Intended use: Research Use Only (RUO)
- R‑phrase: R20, R21, R22
- Storage: Freeze (≤15 °C); minimise light exposure
The fluorescence microscope is configured with a Cy5 filter for both excitation and emission, and the recommended plate for use is a black wall with a clear bottom.
- Beskrivelse:LiveONLY™ Nuclear Red
- Ex/Em:652/682 nm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



