Orders LinkContactUsLinkComponent
Temperature Data Loggers
Temperature Data Loggers
New Product
Leverandør:   RAYPA
Temperature Data Loggers
Leverandør:  RAYPA
New Product
Certifikater
Temperature Data Loggers
Temperature Data Loggers
6 tilgængelige muligheder nedenfor
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
  • User-friendly operation
  • Data logger available in multiple sizes to suit different needs
  • Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Product Family Options
Produktinformation
  • B×D×H
    Tilgængelighed
    Price
  • ESPIBDLDISK_CL
    Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC
    Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC
    60×95×175 mm (External)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product
    New Product
  • ESPIBDL-DISK3618_T
    AISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
    AISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
    60×95×175 mm (External)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product
    New Product
  • ESPIBDLDISK_TL
    Stainless steel and plastic connection base for Communication between the data logger and the software installed on a PC, model: BDLDISK_TL, Weight: 120 g
    Stainless steel and plastic connection base for Communication between the data logger and the software installed on a PC, model: BDLDISK_TL, Weight: 120 g
    60×95×175 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product
    New Product
  • ESPIBDL-DISK3618_C
    AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
    AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
    60×95×175 mm (External)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product
    New Product
  • ESPIDLDISK3618_TL
    AISI-316 L stainless steel disk temperature recorder for Accurate and precise temperature monitoring, Ø×H: 36×18 mm, Model: DLDISK3618_TL, Weight: 80 g
    AISI-316 L stainless steel disk temperature recorder for Accurate and precise temperature monitoring, Ø×H: 36×18 mm, Model: DLDISK3618_TL, Weight: 80 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product
    New Product
  • ESPIDLDISK3618_CL
    AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
    AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
    Specifikationer:
    New Product