Orders LinkContactUsLinkComponent
VWR® Socket Caps and Colour Inserts for Screw Microtubes
VWR® Socket Caps and Colour Inserts for Screw Microtubes
New Product
Leverandør:   VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Leverandør:  VWR Collection
New Product
Certifikater
VWR® Socket Caps and Colour Inserts for Screw Microtubes
VWR® Socket Caps and Colour Inserts for Screw Microtubes
4 tilgængelige muligheder nedenfor
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • 525-1685
    Customisable screw caps, non sterile
    Customisable screw caps, non sterile
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 430,00 kr
    430,00 kr
    1 * 500 Items
       430,00 kr
    Specifikationer:
    New Product
    New Product
  • 525-1649
    Customisable screw caps, non sterile
    Customisable screw caps, non sterile
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 448,00 kr
    448,00 kr
    1 * 500 Items
       448,00 kr
    Specifikationer:
    New Product
    New Product
  • 525-1676
    Customisable screw caps, non sterile
    Customisable screw caps, non sterile
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 448,00 kr
    448,00 kr
    1 * 500 Items
       448,00 kr
    Specifikationer:
    New Product
    New Product
  • 525-1671
    Customisable screw caps, non sterile
    Customisable screw caps, non sterile
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 448,00 kr
    448,00 kr
    1 * 500 Items
       448,00 kr
    Specifikationer:
    New Product