Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Plasmid+® Media

Plasmid+® Media

Plasmid+® media, 1 L bottle
Katalog # THMS446300
Leverandør:   THOMSON INSTRUMENT
Plasmid+® Media
Katalog # THMS446300
Leverandør:  THOMSON INSTRUMENT
CAS nummer:  
Plasmid+® Media
Plasmid+® Media

Plasmid+® media, 1 L bottle

CAS nummer:  
Price
4.180,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Plasmid+® Enriched Media is an optimised microbial growth medium formulated to drive high-copy plasmid production with clean downstreams. When paired with Ultra Yield® Flasks and AirOtop® closures, customers have reported plasmid yield increases of up to 610%.

  • Optimised formulation for E. coli growth and high-copy plasmids

  • Supports low endotoxin output to reduce downstream cleanup
  • Consistent, reproducible results across culture runs
  • Works on standard orbital shakers from bench to pilot volumes

The Ultra Yield® System combines Ultra Yield® Flasks, AirOtop® Enhanced Seals/Vented Caps, and Plasmid+® Enriched Media to boost microbial productivity - especially E. coli - for plasmid DNA and protein expression. Across customer studies, the system has achieved 85%+ supercoiled plasmid DNA with very low endotoxin, delivered 4–10× material cost savings with about 50% less FTE time, and shown roughly 10× higher aeration with smooth scalability from 125 ml to 2.5 L. Bidirectional Transfer Caps on 1.5/2.5 L flasks further streamline sterile harvests or transfers to larger fermenters.

Specifikationer

Specifikationer

  • Beskrivelse:
    Plasmid+® media
  • Størrelse:
    1 L bottle
  • Cat. No.:
    THMS446300
  • Sterility:
    Sterile

Specifikationer