Orders LinkContactUsLinkComponent
Congo Red ≥98% Amyloid fibril-binding dye
Congo Red ≥98% Amyloid fibril-binding dye
Katalog # ABCAAB145645-100
Leverandør:   Abcam
Congo Red ≥98% Amyloid fibril-binding dye
Katalog # ABCAAB145645-100
Leverandør:  Abcam
CAS nummer:  
Congo Red ≥98% Amyloid fibril-binding dye
CAS nummer:  
Price
588,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents
Amyloid fibril-binding dye. Caspase inhibitor. Red shifts on aggregation and binding to amyloid fibrils. Blue shifts on binding to cellulose. Shows apple green birefringence under polarised light in the presence of amyloid fibrils. Inhibits Fib-beta A neurotoxicity.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonymer:  CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
  • Molecular Weight:696,7 g/mol