Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

ec23 ≥98% photostable synthetic retinoid

Marketsource
Leverandør:   Abcam
ec23 ≥98% photostable synthetic retinoid
Leverandør:  Abcam
Marketsource
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents

Photostable synthetic retinoid. All-trans retinoic acid (ATRA) analog. Displays similar biological activity to ATRA. Potently induces stem cell differentiation and enables improved consistency of neuronal differentiation. Induces neurogenesis.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylethynyl) benzoic acid, 4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethynyl)benzoic acid, EC23
  • CAS nummer:104561-41-3
  • Molecular Weight:332,44 g/mol
Specifications for Product Family

Specifikationstestresultater

  • Constituents::
    100% 4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 mg
    1 * 10 mg - 1.260,00 kr
    1.260,00 kr
    1 * 10 mg
       1.260,00 kr

    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 mg
    1 * 50 mg - 5.170,00 kr
    5.170,00 kr
    1 * 50 mg
       5.170,00 kr

    Specifikationer:
    Marketsource
    Marketsource