Orders LinkContactUsLinkComponent
U0126 ≥98% Selective MKK inhibitor
Marketsource
 :   Abcam
U0126 ≥98% Selective MKK inhibitor
 :  Abcam
Marketsource
U0126 ≥98% Selective MKK inhibitor
Selective non-competitive inhibitor of MAP kinase kinase (MKK). Inhibits MKK1, MKK2 (IC₅₀ values of 0,07 and 0,06 μM, respectively) and MKK5 with little or no effect on a wide range of other kinases. Centrally active following systemic administration in vivo.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
  •  :109511-58-2
  •  :380,5 g/mol
  • Constituents::
    100% 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene
Product Family Options
  •  
  •    
    1 * 5 mg
    1 * 5 mg - 1.810,00 kr
    1.810,00 kr
    1 * 5 mg
       1.810,00 kr
    Marketsource
    Marketsource
  •    
    1 * 25 mg
    1 * 25 mg - 7.210,00 kr
    7.210,00 kr
    1 * 25 mg
       7.210,00 kr
    Marketsource
    Marketsource
  •    
    1 * 1 mg
    1 * 1 mg - 745,00 kr
    745,00 kr
    1 * 1 mg
       745,00 kr