Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Fenofibrate ≥99% PPAR-alpha agonist

Fenofibrate ≥99% PPAR-alpha agonist

Katalog # ABCAAB120832-5
Marketsource
Leverandør:   Abcam
Fenofibrate ≥99% PPAR-alpha agonist
Katalog # ABCAAB120832-5
Leverandør:  Abcam
Marketsource
CAS nummer:  49562-28-9
Fenofibrate ≥99% PPAR-alpha agonist
CAS nummer:  49562-28-9
Price
510,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Potent and selective PPAR-α agonist (EC₅₀ values are 18 and 30 μM at mouse and human receptors, respectively). Affinity is 10-fold less at PPAR-γ (EC₅₀ values are 250 and 300 μM at mouse and human receptors, respectively). Hypolipidemic agent.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
  • CAS nummer:49562-28-9
  • Molecular Weight:360,84 g/mol
Specifikationer

Specifikationstestresultater

  • Constituents::
    100% Fenofibrate
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.