Orders LinkContactUsLinkComponent
Polysorbate 80 solution 0,1
Marketsource
Leverandør:   Abcam
Polysorbate 80 solution 0,1
Leverandør:  Abcam
Marketsource
Certifikater
3 tilgængelige muligheder nedenfor
Product Details & Documents
Tween 80 Detergent, 10% Solution, ab291111 (previously known as Tween® 80, MegaPure™ Detergent, 10% Solution, 2126) is a non-ionic detergent useful for the extraction of membrane-bound proteins as well as isolation of nuclei from ma mMalian cell lines.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonymer:  Sorbitanmono(cis-9-octadecenoat), ethoxyliert
  • CAS nummer:9005-65-6
Specifications for Product Family
Specifikationstestresultater
  • Constituents::
    10% Tween 80
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 5.210,00 kr
    5.210,00 kr
    1 * 500 mL
       5.210,00 kr
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 mL
    1 * 50 mL - 745,00 kr
    745,00 kr
    1 * 50 mL
       745,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 1.180,00 kr
    1.180,00 kr
    1 * 100 mL
       1.180,00 kr
    Specifikationer:
    Marketsource