Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cholic acid ≥98% non-denaturing ionic detergent

Cholic acid ≥98% non-denaturing ionic detergent

Katalog # ABCAAB146304-50
Marketsource
Leverandør:   Abcam
Cholic acid ≥98% non-denaturing ionic detergent
Katalog # ABCAAB146304-50
Leverandør:  Abcam
Marketsource
CAS nummer:  81-25-4
Cholic acid ≥98% non-denaturing ionic detergent
CAS nummer:  81-25-4
Price
588,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Non-denaturing ionic detergent used for extraction of membrane proteins. Major primary bile acid produced in the liver and usually conjugated with glycine or taurine to form bile salts. Facilitates fat absorption and cholesterol excretion.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
  • CAS nummer:81-25-4
  • Molecular Weight:408,6 g/mol
Specifikationer

Specifikationstestresultater

  • Constituents::
    100% Cholic acid