Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Lithium chloride ≥99% GSK3beta inhibitor

Marketsource
Leverandør:   Abcam
Lithium chloride ≥99% GSK3beta inhibitor
Leverandør:  Abcam
Marketsource
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents

Direct, non-competitive GSK3β inhibitor in vitro(Ki = 2 mM). Elicits various neuropharmacological effects.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS nummer:7447-41-8
  • Molecular Weight:42,39 g/mol
Specifications for Product Family

Specifikationstestresultater

  • Constituents::
    100% Lithium chloride
Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 g
    1 * 5 g - 666,00 kr
    666,00 kr
    1 * 5 g
       666,00 kr

    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 g
    1 * 1 g - 431,00 kr
    431,00 kr
    1 * 1 g
       431,00 kr

    Specifikationer:
    Marketsource
    Marketsource