CJ-21058 ≥98% SecA inhibitor
SecA inhibitor. Shows antiprotozoal effects (IC₅₀ =7 μM, T. brucei). Inhibits post-translational protein transport into the endoplasmic reticulum. Inhibits ATP-dependant translocation of precursor proteins across a bacterial cell membrane. Shows antibacterial and antifungal effects in vivo.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- :387,5 g/mol
SecA inhibitor. Shows antiprotozoal effects (IC₅₀ =7 μM, T. brucei). Inhibits post-translational protein transport into the endoplasmic reticulum. Inhibits ATP-dependant translocation of precursor proteins across a bacterial cell membrane. Shows antibacterial and antifungal effects in vivo.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- :387,5 g/mol
- Constituents::100% (3E,5S)-3-[[(1R,2S,4aR,6R,8aS)-1,3,6-trimethyl-2-[(E)-prop-1-enyl]-4a,5,6,7,8,8a-hexahydro-2H-naphthalen-1-yl]-hydroxymethylidene]-5-(hydroxymethyl)-1-methylpyrrolidine-2,4-dione
Product Family Options
- MarketsourceMarketsource
unology General
- MarketsourceMarketsource