Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Latrunculin A ≥95% Actin polymerization inhibitor

Marketsource
Leverandør:   Abcam
Latrunculin A ≥95% Actin polymerization inhibitor
Leverandør:  Abcam
Marketsource
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents

Actin polymerization inhibitor. Potent antiproliferative agent. Stabilizes monomeric G-actin. Shows greater potency than Latrunculin B. Binds actin monomers near the nucleotide binding cleft with 1:1 stoichiometry. Inhibits growth of cancer cell lines (IC₅₀ values are 142, 142, 142, 166 and 95 nM for A549, H522-T1, HT-29, U-937 and MDA-MB-43 cells respectively). Shows convulsant effects in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one
  • CAS nummer:76343-93-6
  • Molecular Weight:421,6 g/mol
Specifications for Product Family

Specifikationstestresultater

  • Constituents::
    100% Latrunculin A
Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 1.570,00 kr
    1.570,00 kr
    1 * 1 Each
       1.570,00 kr

    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 6.040,00 kr
    6.040,00 kr
    1 * 1 Each
       6.040,00 kr

    Specifikationer:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.