Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Oil Red O Lysochrome diazo dye

Katalog # ABCAAB146295-50
Marketsource
Leverandør:   Abcam
Oil Red O Lysochrome diazo dye
Katalog # ABCAAB146295-50
Leverandør:  Abcam
Marketsource
CAS nummer:  1320-06-5
Oil Red O Lysochrome diazo dye
CAS nummer:  1320-06-5
Price
kr588.00
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  CI 26125
  • CAS nummer:1320-06-5
  • Molecular Weight:408,51 g/mol
Specifikationer

Specifikationstestresultater

  • Constituents::
    100% 1-[[4-[(dimethylphenyl)azo]dimethylphenyl]azo]-2-naphthol