Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Genistein ≥99% Protein tyrosine kinase inhibitor

Genistein ≥99% Protein tyrosine kinase inhibitor

Marketsource
Leverandør:   Abcam
Genistein ≥99% Protein tyrosine kinase inhibitor
Leverandør:  Abcam
Marketsource
Certifikater
Genistein ≥99% Protein tyrosine kinase inhibitor

3 tilgængelige muligheder nedenfor

Product Details & Documents

Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
  • CAS nummer:446-72-0
  • Molecular Weight:270,24 g/mol
Specifications for Product Family

Specifikationstestresultater

  • Constituents::
    100% Genistein
Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 mg
    1 * 10 mg - 470,00 kr
    470,00 kr
    1 * 10 mg
       470,00 kr

    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 mg
    1 * 50 mg - 1.610,00 kr
    1.610,00 kr
    1 * 50 mg
       1.610,00 kr

    Specifikationer:
    Marketsource
    Marketsource
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mg
    1 * 100 mg - 2.670,00 kr
    2.670,00 kr
    1 * 100 mg
       2.670,00 kr

    Specifikationer:
    Marketsource
    Marketsource