Orders LinkContactUsLinkComponent
Pyronin G Cationic dye (RNA)
Marketsource
 :   Abcam
Pyronin G Cationic dye (RNA)
 :  Abcam
Marketsource
Pyronin G Cationic dye (RNA)
Cationic dye that intercalates RNA and is visible as a red band during electrophoresis. Shown to accumulate in the mitochondria of viable cells. At low concentrations the cytostatic effect is cell arrest in the G1 phase, whereas at high concentrations the cytotoxic effect is cell arrest in the G2 and S phase. Used in combination with Alcian Blue to assess and study pulmonary diseases, and in combination with Methyl Green to stain nucleic acids in paraffin sections. Additionally, has been used to measure RNA in CHO cells by flow cytometry.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
  •  :92-32-0
  •  :302,8 g/mol
  • Constituents::
    100% 3,6-bis(dimethylamino)-10lambda4-xanthen-10-ylium chloride
Product Family Options
eral Biochemicals
  •  
  •    
    1 * 25 g
    1 * 25 g - 2.550,00 kr
    2.550,00 kr
    1 * 25 g
       2.550,00 kr
    Marketsource
    Marketsource
  •    
    1 * 5 g
    1 * 5 g - 705,00 kr
    705,00 kr
    1 * 5 g
       705,00 kr