Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

SICCO Protective Tray

Protective tray, stainless steel welded, 828×26×528 mm
  BOHLV1971-07
 :   Bohlender
SICCO Protective Tray
  BOHLV1971-07
 :  Bohlender
SICCO Protective Tray
SICCO Protective Tray

Protective tray, stainless steel welded, 828×26×528 mm

 
4.160,00 kr
   
1 * 1 Items
 

Suitable for all SICCO glove boxes.

  • Collection tray function: approx. 20 mm height/10 liters

Additional protection for the base panel of the glove box.

 
  • Beskrivelse:
    Protective tray, stainless steel welded, 828×26×528 mm