Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0), Abcam

Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0), Abcam

Leverandør:   Abcam
Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0), Abcam
Leverandør:  Abcam
Certifikater
Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0), Abcam

2 tilgængelige muligheder nedenfor

Product Details & Documents

Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0) is a special solution that enables rehydrate and target retrieval in formalin-fixed, paraffin-embedded tissue sections in one step.

  • pH: 8 Constituents: 5% Tetrasodium ethylene diamine tetraacetate, 1.5% 2-butoxyethanol

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifikationer

Specifikationer

  • Catalog No:
  • ABCAAB93680-125
  • ABCAAB93680-250
  • Opbevaringstemperatur:
  • Store at Ambient.
  • Store at Ambient.

Specifikationer

Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • ABCAAB93680-125
    Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0)
    Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 125 mL
    1 * 125 mL - 2.860,00 kr
    2.860,00 kr
    1 * 125 mL
       2.860,00 kr

    Specifikationer:
    More
    Specifications
    Cat. No.
    ABCAAB93680-125
    Opbevaringstemperatur
    Store at Ambient.
  • ABCAAB93680-250
    Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0)
    Antigen Retrieval Buffer (100X EDTA Buffer, pH 8.0)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 250 mL
    1 * 250 mL - 4.230,00 kr
    4.230,00 kr
    1 * 250 mL
       4.230,00 kr

    Specifikationer:
    More
    Specifications
    Cat. No.
    ABCAAB93680-250
    Opbevaringstemperatur
    Store at Ambient.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.