Orders LinkContactUsLinkComponent
ECL Substrate Kit (High Sensitivity), Abcam
ECL Substrate Kit (High Sensitivity), Abcam
Marketsource
Leverandør:   Abcam
ECL Substrate Kit (High Sensitivity), Abcam
Leverandør:  Abcam
Marketsource
Certifikater
3 tilgængelige muligheder nedenfor
Product Details & Documents
High Sensitivity ECL Substrate Kit ab133406 is designed for the detection of proteins with 23pg-187ng of protein per band.
Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • ABCAAB133406-200
  • ABCAAB133406-20
  • ABCAAB133406-500
  • Opbevaringstemperatur:
  • Store at +4°C.
  • Store at +4°C.
  • Store at +4°C.
Specifikationer
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • ABCAAB133406-200
    ECL Substrate Kit (High Sensitivity)
    ECL Substrate Kit (High Sensitivity)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 200 mL
    1 * 200 mL - 2.590,00 kr
    2.590,00 kr
    1 * 200 mL
       2.590,00 kr
    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB133406-200
    Opbevaringstemperatur
    Store at +4°C.
    Marketsource
  • ABCAAB133406-20
    ECL Substrate Kit (High Sensitivity)
    ECL Substrate Kit (High Sensitivity)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 mL
    1 * 20 mL - 784,00 kr
    784,00 kr
    1 * 20 mL
       784,00 kr
    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB133406-20
    Opbevaringstemperatur
    Store at +4°C.
    Marketsource
unology General
Produktinformation
  • Tilgængelighed
    Price
  • ABCAAB133406-500
    ECL Substrate Kit (High Sensitivity)
    ECL Substrate Kit (High Sensitivity)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 4.550,00 kr
    4.550,00 kr
    1 * 500 mL
       4.550,00 kr
    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB133406-500
    Opbevaringstemperatur
    Store at +4°C.
    Marketsource