Orders LinkContactUsLinkComponent
RNase R
RNase R
Katalog # ABCAAB286929-1KIT
Marketsource
Leverandør:   Abcam
RNase R
Katalog # ABCAAB286929-1KIT
Leverandør:  Abcam
Marketsource
CAS nummer:  
CAS nummer:  
Price
2.820,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
RNase R is an E. coli exoribonuclease that exhibits 3'-to-5' exonuclease activity, efficiently digesting nearly all linear RNA species. - Efficiently digests linear RNA species with 3'-to-5' exonuclease activity - High purity – contains RNase R at 10 U/µl for optimal performance
Advarsel: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
  • Cat. No.:
    ABCAAB286929-1KIT
  • Enzym navn:
    RNase R
Specifikationer