Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lanreotide

Lanreotide

Marketsource
Leverandør:   Abcam
Lanreotide
Leverandør:  Abcam
Marketsource
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents

MW 1096.3 Da, Purity >95%. Lanreotide, Somatostatin receptor analog. Achieve your results faster with highly validated, pure and trusted compounds.

  • Constituents: 100% [cyclo S-S]-3-(2- naphthyl)-D-alanyl-L-cysteinyl-L-tryrosyl-D-tryptophyl-L-lysyl-L-valyl-L- cysteinyl-L-threoninamide

Advarsel: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifikationer

Specifikationer

  • Catalog No:
  • ABCAAB141209-1
  • ABCAAB141209-5
  • Protein/peptide name:
  • Lanreotide
  • Lanreotide
  • CAS-nr:
  • 1 mg
  • Renhed:
  • >95%
  • >95%
  • Molekylvægt:
  • 1096.3 DA
  • 1096.3 DA
  • Opbevaring betingelser:
  • Store at -20 °C.
  • Store at -20 °C.
  • Shipping temperature:
  • Ambient - Can Ship with Ice
  • Ambient - Can Ship with Ice

Specifikationer

Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 mg
    1 * 1 mg - 510,00 kr
    510,00 kr
    1 * 1 mg
       510,00 kr

    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB141209-1
    Protein/peptide name
    Lanreotide
    CAS-nr
    1 mg
    Renhed
    >95%
    Molekylvægt
    1096.3 DA
    Opbevaring betingelser
    Store at -20 °C.
    Shipping temperature
    Ambient - Can Ship with Ice
    Marketsource

unology General

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 mg
    1 * 5 mg - 823,00 kr
    823,00 kr
    1 * 5 mg
       823,00 kr

    Specifikationer:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB141209-5
    Protein/peptide name
    Lanreotide
    CAS-nr
    Renhed
    >95%
    Molekylvægt
    1096.3 DA
    Opbevaring betingelser
    Store at -20 °C.
    Shipping temperature
    Ambient - Can Ship with Ice
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.