Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Endomorphin-1

Endomorphin-1

Katalog # ABCAAB120411-5
Marketsource
Leverandør:   Abcam
Endomorphin-1
Katalog # ABCAAB120411-5
Leverandør:  Abcam
Marketsource
CAS nummer:  
Endomorphin-1
CAS nummer:  
Price
862,00 kr
Log ind for tilgængelighed
1 * 5 mg
Product Details & Documents

MW 610.67 g/mol, Purity >95%. Endogenous peptide with extremely high affinity for μ opioid receptors (Ki = 360 pM). Displays 4000- and 15000-fold selectivity over δ and κ receptors, respectively.

  • Constituents: 100% Endomorphin-1

Advarsel: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Cat. No.:
    ABCAAB120411-5
  • Protein/peptide name:
    Endomorphin-1
  • Renhed:
    >95%
  • Opbevaring betingelser:
    Store at -20 °C.
  • Shipping temperature:
    Ambient - Can Ship with Ice

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.