Orders LinkContactUsLinkComponent
G-Biosciences TAE Running Buffer (50X)
G-Biosciences TAE Running Buffer (50X)
Leverandør:   G-Biosciences
G-Biosciences TAE Running Buffer (50X)
Leverandør:  G-Biosciences
Certifikater
G-Biosciences TAE Running Buffer (50X)
2 tilgængelige muligheder nedenfor
Product Details & Documents
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
  • 50X Concentrated solution: 2 M Tris-Acetate, 0,05 M EDTA (pH 8,3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.
Product Family Options
Produktinformation
  • Pakning
    Pakn
    Tilgængelighed
    Price
  • R023
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
    Plastic bottle
    1 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 L
    1 * 1 L - 721,00 kr
    721,00 kr
    1 * 1 L
       721,00 kr
  • R024
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
    4 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 L
    1 * 4 L - 1.850,00 kr
    1.850,00 kr
    1 * 4 L
       1.850,00 kr