Orders LinkContactUsLinkComponent
G-Biosciences Acetate Buffer
G-Biosciences Acetate Buffer
Leverandør:   G-Biosciences
G-Biosciences Acetate Buffer
Leverandør:  G-Biosciences
Certifikater
G-Biosciences Acetate Buffer
2 tilgængelige muligheder nedenfor
Product Details & Documents
Acetate buffer at pH 5,0 and 5,5 are supplied as ready to use solutions that eliminate the calculation, formulation and weighing errors.
  • Precision in pH
  • Guaranteed reproducibility
Acetate buffer, pH 5,0 buffer is commonly used in protein purification, enzyme stabilisation, and histology.
Acetate buffer, pH 5,5 is useful for preserving enzyme activity or biomolecules that are sensitive to lower pH.
Product Family Options
eral Biochemicals
Produktinformation
  • Volumen
    Tilgængelighed
    Price
  • GENO786-876
    Acetate buffer, pH 5,0 (200 mM)
    Acetate buffer, pH 5,0 (200 mM)
    250 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 250 mL
    1 * 250 mL - 532,00 kr
    532,00 kr
    1 * 250 mL
       532,00 kr
  • GENO786-877
    Acetate buffer, pH 5,5 (200 mM)
    Acetate buffer, pH 5,5 (200 mM)
    250 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 250 mL
    1 * 250 mL - 532,00 kr
    532,00 kr
    1 * 250 mL
       532,00 kr